Learn More
Mouse anti-CD96, Clone: MM0690-1H29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236784
Catalog No. NBP236784
Add to cart
Description
Description
CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CD96
MM0690-1H29
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
CD96
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Recombinant human CD96 V2 isoform.
Protein G purified
RUO
Primary
10225
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Western Blot
LYOPH
Western Blot 1:500-1000
P40200
CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Mouse
IgG2
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes human CD96 V2 isoform by direct ELISA and WB.
Human