Learn More
Mouse anti-CD98, Alexa Fluor 647, Clone: 1C11.7E3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236491AF647
Catalog No. NBP236491Y
Add to cart
Description
Description
CD98 Monoclonal antibody specifically detects CD98 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD98
1C11.7E3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
P08195
4F2, 4F2hc, 4T2HC, antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43,4F2HC, CD98, CD98 antigen, CD98 heavy chain, CD98HC, heavy chain, Lymphocyte activation antigen 4F2 large subunit, MDU1antigen defined by monoclonal 4F2, heavy chain, monoclonal 44D7, NACAE4F2 cell-surface antigen heavy chain, solute carrier family 3 (activators of dibasic and neutral amino acidtransport), member 2,4F2 heavy chain antigen
Mouse
IgG2b Kappa
0.1 ml
Cell Biology, Cellular Markers, Immunology, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary
6520
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SLC3A2
Partial recominant human CD98 (between amino acids 200-400) [UniProt P08195]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human