Learn More
Mouse anti-CEACAM-5/CD66e, Clone: CB30, Alexa Fluor 405, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11058734AF405
Catalog No. NB11058734W
Add to cart
Description
Description
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CEACAM5/CD66e
CB30
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CEACAM5
Human carcinoembryonic antigen (CEA; CEACAM5)
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The antibody CB30 recognizes CD66e (CEA; 180-200 kDa), a cell surface bound carcinoembryonic antigen mainly expressed on epithelial cells.
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
IgG1
0.1 ml
Cancer, Cellular Markers, Immunology
Primary
1048
Protein A purified