Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centrin 1 Antibody (147), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP243803

Catalog No. NBP243803


Add to Cart

Description

Description

Ensure accurate, reproducible results in Western Blot

Centrin 1 Monoclonal specifically detects Centrin 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Centrin 1
Monoclonal
Unconjugated
PBS with No Preservative
CETN1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Centrin 1. The exact sequence is proprietary.
Protein G purified
RUO
Primary
Human
Purified
Western Blot
147
Western Blot 1:500-1:3000
calcium binding protein, caltractin, Caltractin isoform 2, CEN1centrin-1, centrin, EF-hand protein, 1, CETN, EF-hand protein
Mouse
20 kDa
0.1 mL
Cell Cycle and Replication
1068
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only