Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHFR, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant CHFR.

Manufacturer:  Abnova Corporation H00055743A01

Catalog No. 89-107-060


Add to cart

Description

Description

Sequence: MERPEEGKQSPPPQPWGRLLRLGAEEGEPHVLLRKREWTIGRRRGCDLSFPSNKLVSGDHCRIVVDEKSGQVTLEDTSTSGTVINKLKVVKKQTCPLQTGDVIYLVYRKNEPEHNVAYLYESLSEKQGMTQESFDTSGAGAGRGADPRVPPSSPATQVCFEEPQPSTSTSDLFPTASASSTEPSPAGRERSSSCGSGGGGISPKGSGPSVASDEVSSFASALPDRKTASFSSLEPQDQEDLEPVKKKMRGDGDLDLNGQLLVAQPRRNAQTVHEDVRAAAGKPDKMEETLTCIICQDLLHDCVSLQPCMHTFCAACYSGWMERSSLCPTCRCPVERICKNHILNNLVEAYLIQHPDKSRSEEDVQSMDARNKITQDMLQPKVRRSFSDEEGSSEDLLELSDVDSESSDISQPYVVCRQCPEYRRQAAQPPHCPAPEGEPGAPQALGDAPSTSVSLTTAVQDYVCPLQGSHALCTCCFQPMPDRRVEREQDPRVAPQQCAVCLQPFCHLYWGCTRTGCYGCLAPFCELNLGDKCLDGVLNNNSYESDILKNYLATRGLTWKNMLTESLVALQRGVFLLSDYRVTGDTVLCYCCGLRSFRELTYQYRQNIPASELPVAVTSRPDCYWGRNCRTQVKAHHAMKFNHICEQTRFKN
Specifications

Specifications

CHFR
Unconjugated
50% glycerol
BC012072
FLJ10796/FLJ33629/RNF116/RNF196
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA
Mouse polyclonal antibody raised against a full-length recombinant CHFR.
CHFR
Antisera
CHFR
CHFR (AAH12072.1, 1 a.a. to 652 a.a) full-length recombinant protein with GST tag.
RUO
Primary
55743
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only