Learn More
Chymotrypsin, Mouse, Clone: 602, Abnova™
Mouse monoclonal antibody raised against native Chymotrypsin.
Manufacturer:  Abnova Corporation MAB4163
Catalog No. 89-135-634
Add to cart
Description
Description
Specifications
Specifications
Chymotrypsin
602
Mouse monoclonal antibody raised against native Chymotrypsin.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG3, kappa
RUO
Primary
Does not cross-react with human pancreatic trypsin.
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native purified Chymotrypsin from human pancreatic.
200 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human