Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-cleavage stimulation factor, Clone: 10E11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242659

Catalog No. NBP242659


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

cleavage stimulation factor Monoclonal antibody specifically detects cleavage stimulation factor in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

cleavage stimulation factor
1000000000000
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Purified
CSTF1
Recombinant human CSTF1 (1-431aa) purified from E. coli
Protein A purified
RUO
Primary
1477
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
NP_001028693
CF-1 50 kDa subunit, Cleavage stimulation factor 50 kDa subunit, cleavage stimulation factor subunit 1, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD, cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa, CSTF 50 kDa subunit, CstF-50, CstFp50
Mouse
IgG2b Kappa
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only