Learn More
COL3A1, Mouse, Clone: COL-29, Abnova™
Mouse monoclonal antibody raised against native COL3A1.
Supplier: Abnova Corporation MAB1514
Description
This gene encodes the pro-alpha1 chains of type III collagen, a fibrillar collagen that is found in extensible connective tissues such as skin, lung, uterus, intestine and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome types IV, and with aortic and arterial aneurysms. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene. [provided by R. Dalgleish
Specifications
COL3A1 | |
Monoclonal | |
Unconjugated | |
Western Blot (0.25-0.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Immunohistochemistry (Frozen sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
COL3A1 | |
COL3A1 | |
Native purified human COL3A1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Immunohistochemistry, Western Blot | |
FH-7A | |
Mouse monoclonal antibody raised against native COL3A1. | |
Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
EDS4A/FLJ34534 | |
Mouse | |
Affinity purification | |
RUO | |
1281 | |
Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.