Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Complement 4d, Clone: C4D203, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2447940.02MG

 View more versions of this product

Catalog No. NBP24479400


Add to cart

Description

Description

Ensure accurate, reproducible results in ELISA, Immunofluorescence

Complement C4b/d Monoclonal antibody specifically detects Complement C4b/d in Human samples. It is validated for SDS-Page, Immunofluorescence.
Specifications

Specifications

Complement 4d
C4D203
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
C4d
Mouse
IgG1
Protein A or G purified
RUO
Primary
This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.C4d is a degradation product of the activated complement factor C4b. Complement 4b is typically activated by binding of Abs to specific target molecules. Following activation and degradation of the C4 molecule, thio-ester groups are exposed, which allow transient, covalent binding of the degradation product Complement 4d to endothelial cell surfaces and extracellular matrix components of vascular basement membranes near the sites of C4 activation. The presence of C4d in peritubular capillaries is a key indicator for acute humoral (i.e. antibody-mediated) rejection of kidney, heart, pancreas and lung allografts. As an established marker of antibody-mediated acute renal allograft rejection and its proclivity for endothelium, this component can be detected in peritubular capillaries in chronic renal allograft rejection as well as hyperacute rejection, acute vascular rejection, acute cellular rejection, and borderline rejection. It has been shown to be a significant predictor of transplant kidney graft survival. Anti-C4d, combined with anti-C3d, can be utilized as a tool for diagnosis of allograft rejection that may warrant a prompt and aggressive anti-rejection treatment.
SDS-Page, Immunofluorescence
0.2 mg/mL
SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml
Purified
C4A
Recombinant human Complement 4d protein
192 kDa
0.02 mg
Store at 4C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only