Learn More
Mouse anti-Complement 4d, Clone: TDM16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2447930.2MG
 View more versions of this product
Catalog No. NBP24479302
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ensure accurate, reproducible results in ELISA, Immunofluorescence
Specifications
Specifications
Complement 4d
TDM16
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
C4d
Mouse
IgG1
Protein A or G purified
RUO
Primary
This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.C4d is a degradation product of the activated complement factor C4b. Complement 4b is typically activated by binding of Abs to specific target molecules. Following activation and degradation of the C4 molecule, thio-ester groups are exposed, which allow transient, covalent binding of the degradation product Complement 4d to endothelial cell surfaces and extracellular matrix components of vascular basement membranes near the sites of C4 activation. The presence of C4d in peritubular capillaries is a key indicator for acute humoral (i.e. antibody-mediated) rejection of kidney, heart, pancreas and lung allografts. As an established marker of antibody-mediated acute renal allograft rejection and its proclivity for endothelium, this component can be detected in peritubular capillaries in chronic renal allograft rejection as well as hyperacute rejection, acute vascular rejection, acute cellular rejection, and borderline rejection. It has been shown to be a significant predictor of transplant kidney graft survival. Anti-C4d, combined with anti-C3d, can be utilized as a tool for diagnosis of allograft rejection that may warrant a prompt and aggressive anti-rejection treatment.
Immunofluorescence
0.2 mg/mL
Immunofluorescence 0.5 - 1.0 ug/ml
Purified
C4A
Human Complement 4d protein
192 kDa
0.2 mg
Store at 4C.
Monoclonal
Human