Learn More
Cortisol, Mouse, Clone: XM210, Abnova™
Mouse monoclonal antibody raised against Cortisol.
Manufacturer:  Abnova Corporation MAB2801
Catalog No. 89-135-238
Add to cart
Description
Description
Specifications
Specifications
Cortisol
XM210
Mouse monoclonal antibody raised against Cortisol.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG2a
RUO
Primary
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Cortisol conjugated with BSA.
200 ug
Store at 4°C.
Monoclonal