Learn More
Mouse anti-CREB (p Ser133), Clone: MM0704-13B29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236785
Catalog No. NBP236785
Add to cart
Description
Description
CREB p Ser133 Monoclonal antibody specifically detects CREB p Ser133 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CREB (p Ser133)
MM0704-13B29
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
CREB1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing S133 site of human CREB).
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes human phosphorylated CREB S133 in WB.
Human
Immunocytochemistry, Immunofluorescence, Western Blot
LYOPH
Western Blot 1:500-1000, Immunocytochemistry/Immunofluorescence 1:50-100
P16220
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Mouse
IgG2
0.1 mg
Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary
1385
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.