Learn More
Mouse anti-CrkL (p Tyr207), Clone: MM0707-9F39, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236786
Catalog No. NBP236786
Add to cart
Description
Description
CrkL p Tyr207 Monoclonal antibody specifically detects CrkL p Tyr207 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CrkL (p Tyr207)
MM0707-9F39
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
CRKL
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing human CrkL Y207).
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes human CrkL with phospho-Y207 in direct ELISAs and Western blots.
Human
Western Blot
LYOPH
Western Blot 1:500-1000
P46109
crk-like protein, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Mouse
IgG1
0.1 mg
Cancer, Phospho Specific, Signal Transduction
Primary
1399
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.