Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Cytokeratin 17, Clone: 9F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242619

Catalog No. NBP242619


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence

Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Cytokeratin 17
9F3
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Purified
KRT17
Recombinant human KRT17(1-432aa) purified from E. coli
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot, Flow Cytometry 2-5ug, ELISA, Immunocytochemistry/Immunofluorescence 1:200
NP_000413
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Mouse
IgG2a Kappa
100 ul
Cancer
Primary
3872
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only