Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

MilliporeSigma™ Mouse anti-Cytokeratin 5,6, Clone: D5/16B4, Alexa Fluor 647,

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma™ MAB1620AF647

Catalog No. MAB1620AF64



Description

Description

Specifically detects Cytokeratin 5,6 clone: D5/16B4 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

Keratin, type II cytoskeletal 5 (UniProt P13647; also known as 58kDa cytokeratin, CK-5, Cytokeratin-5, K5, Keratin-5, Type-II keratin Kb5) is encoded by the KRT5 (also known as DDD1, DM-EBS, EBSMCE, K-EBS, MP-EBS, WC-EBS) gene (Gene ID 3852) in human. Cytokeratin-5 (Keratin-5 or K5) belongs to a family of highly conserved epithelial-specific intermediate filament (IF) proteins. Each epithelium is characterized by its unique keratin expression pattern. In interfollicular epidermis, four major keratins are synthesized and co-polymerize in pairs, K5/K14 and K1/KI0, to form filaments. The expression of these two pairs is strictly regulated during normal keratinocyte differentiation. The K5/K14 filament is expressed in proliferating basal cells, while the K1/K10 filament is expressed during the transition to terminal differentiation and at the cessation of cell division. Similar to other IF members, Keratin-5 contains a central rod region composed of alpha helix segments (a.a. 168-203, 223-315, 339-477) that allow two different IF proteins, one type I (acidic) and one type II (basic) keratin, to intertwine into a coiled-coil formation via hydrophobic interaction. Kearain-5 is a type II keratin that pairs with the type I keratin-14 to form IF.
Specifications

Specifications

Cytokeratin 5,6
D5/16B4
Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Purified
Murine
IgG1
100μL
Cell Structure
Primary
NM_000424.3
Immunocytochemistry
Alexa Fluor 647
P13647
KRT5; DDD1; DM-EBS; EBSMCE; K-EBS; MP-EBS; WC-EBS
D.
Protein A Purfied
RUO
Stable for one year at 2°C to 8°C from date of receipt.
Monoclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only