Learn More
Mouse anti-DC-SIGN/CD209, Clone: 102E11.06, Alexa Fluor 647, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0202A6471000.05MG
Catalog No. DDX0202Y5MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human, Mouse, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
DC-SIGN/CD209
102E11.06
Alexa Fluor 647
PBS and 50% Glycerol with No Preservative
Purified
CD209
DC-SIGN/CD209 was used as immunogen.
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Canine
Flow Cytometry, Immunohistochemistry
0.5mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry
NP_066978
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Mouse
IgG2b Kappa
0.05 mg
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
30835