Learn More
Mouse anti-DCIR/CLEC4A, Clone: 111F8.04, Alexa Fluor 488, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0180A4881000.05MG
Catalog No. DDX0180X5MG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
Specifications
DCIR/CLEC4A
111F8.04
Alexa Fluor 488
PBS and 50% Glycerol with No Preservative
CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Mouse
IgG1
0.05 mg
Immunology, Innate Immunity
Primary
50856
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Purified
CLEC4A
DCIR was used as immunogen.
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human