Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Decorin, Clone: 1G4C5, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237336

Catalog No. NBP237336

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Decorin
1G4C5
Unconjugated
PBS with 0.05% Sodium Azide
Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B
Mouse
IgG1
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ELISA
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
Affinity Purified
DCN
Purified recombinant fragment of human DCN (AA: 263-324) expressed in E. Coli.
39.7 kDa
0.1 ml
Cancer
Primary
1634
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only