Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody (231), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243570
Description
Ensure accurate, reproducible results in Western Blot, Immunoprecipitation
DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunoprecipitation.Specifications
DYKDDDDK Epitope Tag | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
Mouse | |
1.01 kDa | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Western Blot, Immunoprecipitation | |
231 | |
Western Blot 1:1000-1:10000, Immunoprecipitation 1:100-1:500 | |
DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
The immunogen used to generate this antibody corresponds to DDDDK tag | |
Protein G purified | |
RUO | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction