Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-E-Cadherin, Clone: CDH1/1121 + CDH1/1122, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen E-Cadherin
Clone CDH1/1121 + CDH1/1122
Concentration 0.2mg/mL
Immunogen Recombinant fragment (119 Amino acid residues around aa 400-500) of human E-Cadherin protein
Gene ID (Entrez) 999
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24472002 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2447200.2MG
0.2 mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Specifications

E-Cadherin
0.2mg/mL
999
Monoclonal
Mouse
IgG
Purified
RUO
Protein A or G purified
CDH1/1121 + CDH1/1122
Recombinant fragment (119 Amino acid residues around aa 400-500) of human E-Cadherin protein
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Primary
Recognizes a protein of 120-80kDa, identified as E-cadherin. Cadherins comprise a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of tissue structure and morphogenesis. The classical cadherins, E-, N- and P-cadherin, consist of large extracellular domains characterized by a series of five homologous NH2 terminal repeats. The relatively short intracellular domains interact with a variety of cytoplasmic proteins, such as catenin, to regulate cadherin function. E-cadherin plays an important role in epithelial cell adhesion. A decreased expression of E-cadherin is associated with metastatic potential and poor prognosis in breast cancer, prostate and esophageal cancer. In combination with p120 Catenin, it is useful for the differentiation between ductal (E-cadherin +) and lobular (E-cadherin -) breast carcinomas. It may also help in diagnosis of mesothelioma.
Unconjugated
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Store at 4C.

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit