Learn More
Mouse anti-EEA1, DyLight 680, Clone: 6D4.1D4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236568FR
Catalog No. NBP236568FR
Add to cart
Description
Description
EEA1 Monoclonal antibody specifically detects EEA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
Specifications
EEA1
6D4.1D4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular)
Q15075
early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2
Mouse
IgG1 Kappa
0.1 ml
Endosome Markers, Membrane Trafficking and Chaperones
Primary
8411
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Purified
EEA1
Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse