Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-EEA1, PE, Clone: 6D4.1D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236568PE

Catalog No. NBP236568PE


Add to cart

Description

Description

EEA1 Monoclonal antibody specifically detects EEA1 in Human, Mouse samples. It is validated for Flow (Intracellular).
Specifications

Specifications

EEA1
6D4.1D4
Flow (Intracellular)
Q15075
early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2
Mouse
IgG1 Kappa
0.1 ml
Endosome Markers, Membrane Trafficking and Chaperones
Primary
8411
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
Purified
EEA1
Partial recombinant human EEA1 protein (between amino acids 50-300). [UniProt Q15075]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit