Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-EGF R/ErbB1, Clone: H9B4, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | EGFR |
---|---|
Clone | H9B4 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-1.0ug/ml, ELISA 1-5ug/ml for coating, Immunoprecipitation 1-2ug/500ug protein lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23462802
|
Novus Biologicals
NBP2346280.2 MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
EGFR Monoclonal antibody specifically detects EGFR in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
EGFR | |
1.0 mg/mL | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
10mM PBS - BSA free with No Preservative | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
EGFR | |
IgG1 | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
170 kDa |
H9B4 | |
Western Blot 0.5-1.0ug/ml, ELISA 1-5ug/ml for coating, Immunoprecipitation 1-2ug/500ug protein lysate | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse (Negative), Rat (Negative) | |
P00533 | |
1956 | |
Purified EGFR from A431 cells. | |
Primary | |
Protein A purified | |
This MAb reacts with a cytoplasmic domain of EGFR. EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, beta cellulin & epiregulin. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title