Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2AK4, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant EIF2AK4.

Manufacturer:  Abnova Corporation H00440275A01

Catalog No. 89-107-806


Add to cart

Description

Description

EIF2AK4 belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2S1; MIM 603907) to downregulate protein synthesis in response to varied cellular stresses (Berlanga et al., 1999 [PubMed 10504407]).[supplied by OMIM

Sequence: VVSVGQMSMSRAINLTQKLWTAGITAEIMYDWSQSQEELQEYCRHHEITYVALVSDKEGSHVKVKSFEKERQTEKRVLETELVDHVLQKLRTKVTDERNG
Specifications

Specifications

EIF2AK4
Unconjugated
50% glycerol
NM_001013703
GCN2/KIAA1338
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant EIF2AK4.
EIF2AK4
Antisera
EIF2AK4
EIF2AK4 (NP_001013725, 1401 a.a. to 1500 a.a) partial recombinant protein with GST tag.
RUO
Primary
440275
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only