Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Enterovirus, Clone: 4A7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242840

Catalog No. NBP242840


Add to cart

Description

Description

Ensure accurate, reproducible results in ELISA, Immunocytochemistry/Immunofluorescence

Enterovirus Monoclonal antibody specifically detects Enterovirus in Viral samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Enterovirus
4A7
Unconjugated
0.05M PBS (pH 7.4) with 0.1% Sodium Azide
Mouse
Immunogen affinity purified
RUO
Primary
Virus
ELISA, Immunocytochemistry, Immunofluorescence
3.97 mg/mL
ELISA, Immunocytochemistry/Immunofluorescence
Purified
Coxsakieviruses B5
100 ug
Store at 4C. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only