Learn More
Esrp1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against full-length recombinant Esrp1.
Manufacturer: Abnova Corporation PAB16900
Specifications
Esrp1 | |
Polyclonal | |
Mouse polyclonal antibody raised against full length recombinant Esrp1. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Rbm35a/BC031468/A630065D16/RP23-322G9.1/2210008M09Rik | |
Mouse | |
100 μg | |
Primary | |
Mouse |
Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Esrp1 | |
Esrp1 | |
Recombinant protein corresponding to full length mouse Esrp1. | |
RUO | |
207920 | |
Liquid |