Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBL, Mouse, Clone: 38F3, Abnova™

Mouse monoclonal antibody raised against native FBL.

Manufacturer:  Abnova Corporation MAB2309

Catalog No. 89-135-163


Add to cart

Description

Description

Specifications

Specifications

FBL
38F3
Mouse monoclonal antibody raised against native FBL.
In tissue culture supernatant (10mM sodium azide)
Mouse
IgG1
RUO
Primary
Detects a band of approximately 34 KDa and shows a clear and strong punctate staining of yeast nuclei.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native purified yeast FBL.
250 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Drosophila, Human, Nematode, Plant, Rat, Yeast
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit