Learn More
Mouse anti-Fc epsilon RI, Clone: AER-37, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1432780.025MG
 View more versions of this product
Catalog No. NBP14327803
Add to cart
Description
Description
Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Fc epsilon RI
AER-37
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Mouse
IgG2b Kappa
0.025 mg
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary
2205
Flow Cytometry, Immunohistochemistry (Paraffin)
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry-Paraffin
Affinity Purified
FCER1A
The immunogen for this antibody was FceRI alpha.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human