Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ferritin, Mouse, Clone: 4C11B10, Abnova™
Mouse monoclonal antibody raised against full-length native ferritin.
Supplier: Abnova Corporation MAB1567
Specifications
Ferritin | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
MGC71996 | |
Mouse | |
1 mg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA | |
4C11B10 | |
Mouse monoclonal antibody raised against full length native ferritin. | |
In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide) | |
FTL | |
Native purified ferritin from human liver. | |
RUO | |
2512/2495 | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction