Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-FGF-4, Clone: 2D7D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237375

Catalog No. NBP237375


Add to cart

Description

Description

FGF-4 Monoclonal antibody specifically detects FGF-4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

FGF-4
2D7D5
Western Blot, Flow Cytometry, ELISA 1:10000
Ascites
FGF4
Purified recombinant fragment of human FGF4 (AA: 62-123) expressed in E. Coli.
22 kDa
0.1 ml
Angiogenesis, Biologically Active Proteins, Cancer
Primary
2249
Western Blot, Flow Cytometry, ELISA
Unconjugated
Ascites with 0.03% Sodium Azide
fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only