Learn More
Mouse anti-FHL2, Clone: 11-134, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1545770.05MG
Catalog No. NBP15457705
Add to cart
Description
Description
FHL2 Monoclonal antibody specifically detects FHL2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
FHL2
11-134
Unconjugated
PBS (pH 7.2) and 50% Glycerol with No Preservative
Purified
FHL2
This antibody was purified from hybridoma (clone 11-134) supernatant using protein-A Sepharose. This hybridoma was established by fusion of mouse myeloma cell SP2/0-Ag4 with Balb/c mouse splenocyte immunized with His-FHL2.
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Protein A purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1-5 ug/ml, Immunohistochemistry 1-5 ug/ml, Immunoprecipitation 3 ug/200-300 ul of cell extract, Immunohistochemistry-Paraffin 1-5 ug/ml
Q14192
down-regulated in rhabdomyosarcoma LIM protein, DRALAAG11, FHL-2, four and a half LIM domains 2, four and a half LIM domains protein 2, four and a half LIM-domain protein 2, LIM domain protein DRAL, Skeletal muscle LIM-protein 3, SLIM-3, SLIM3aging-associated gene 11
Mouse
IgG2a
0.05 mg
Angiogenesis, Cancer
Primary
2274
Human, Mouse