Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ38725, Mouse, Clone: 2F7-1D6, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant FLJ38725.

Manufacturer:  Abnova Corporation H00144811M01A

Catalog No. 89-107-639


Add to cart

Description

Description

Sequence: MAEAVLIDLFGLKLNSQKNCHQTLLKTLNAVQYHHAAKAKFLCIMCCSNISYERDGEQDNCEIETSNGLSALLEEFEIVSCPSMAATLYTIKQKIDEKNLSSIKVIVPRHRKTLMKAFIDQLFTDVYNFEFEDLQVTFRGGLFKQSIEINVITAQELRGIQNEIETFLRSLPALRGKLTIITSSLIPDIFIHGFTTRTGGISYIPTLSSFNLFSSSKRRDPKVVVQENLRRLANAAGFNVEKFYRIKTHHSNDIWIMGRKEPDSYDGITTNQRGVTIAALGADCIPIVFADPVKKACGVAHAGWKGTLLGVAMATVNAMIAEYGCSLEDIVVVLGPSVGPCCFTLPRESAEAFHNLHPACVQLFDSPNPCIDIRKATRILLEQGGILPQNIQDQNQDLNLCTSCHPDKFFSHVRDGLNFGTQIGFISIKE
Specifications

Specifications

FLJ38725
Monoclonal
Unconjugated
ascites with no preservative
BC035749
DKFZp686D11119/FLJ38725
Mouse
IgM Kappa
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
2F7-1D6
Mouse monoclonal antibody raised against a full-length recombinant FLJ38725.
C13orf31
Ascites
C13orf31
FLJ38725 (AAH35749,1 a.a. ∽ 430 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
200 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
144811
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit