Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (7E8.2C8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242210FR
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Flt-3/Flk-2/CD135 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7E8.2C8 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P36888 | |
FLT3 | |
Partial recombinant human FLT3 protein (between amino acids 250-500) [UniProt P36888] | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific | |
2322 | |
Store at 4C in the dark. | |
IgG2b λ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction