Learn More
FSHR Mouse anti-Human, DyLight 350, Clone: 6E8.2F5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236489UV
Catalog No. NBP236489UV
Add to cart
Description
Description
FSHR Monoclonal antibody specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
FSHR
6E8.2F5
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
P23945
follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Mouse
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Purified
FSHR
A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945]
0.1 ml
GPCR
Primary
2492