Learn More
Mouse anti-Fumarase, Clone: 7612, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243578
Catalog No. NBP243578
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
Fumarase Monoclonal antibody specifically detects Fumarase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Fumarase
7612
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Purified
FH
Recombinant protein encompassing a sequence within the center region of human Fumarate hydratase. The exact sequence is proprietary.
55 kDa
0.1 ml
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
2271
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS with No Preservative
EC 4.2.1.2, Fumarase, fumarate hydratase, fumarate hydratase, mitochondrial, HLRCC, LRCC, MCL, MCUL1
Mouse
IgG3
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat