Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEF-H1 Antibody (1132), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243584
Description
Ensure accurate, reproducible results in Western Blot
GEF-H1 Monoclonal specifically detects GEF-H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
GEF-H1 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
ARHGEF2 | |
Recombinant protein encompassing a sequence within the C-terminus region of human GEF-H1. The exact sequence is proprietary. | |
0.1 mL | |
Cell Cycle and Replication | |
9181 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin) | |
1132 | |
Western Blot 1:500-1:3000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
GEF, GEFH1, GEF-H1DKFZp547P1516, Guanine nucleotide exchange factor H1, KIAA0651DKFZp547L106, LFP40FLJ45440, Microtubule-regulated Rho-GEF, P40, Proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2, Rho/Rac guanine nucleotide exchange factor (GEF) 2, Rho/Rac guanine nucleotide exchange factor 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction