Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutaminase Antibody (3211), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243762
Description
Ensure accurate, reproducible results in Western Blot
Glutaminase Monoclonal specifically detects Glutaminase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Glutaminase | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
GLS | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Glutaminase C (GAC). The exact sequence is proprietary. | |
Protein G purified | |
RUO | |
2744 | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
3211 | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
AAD20, EC 3.5.1.2, GLS1DKFZp686O15119, glutaminase, glutaminase C, glutaminase kidney isoform, mitochondrial, glutaminase, phosphate-activated, K-glutaminase, KIAA0838FLJ10358, L-glutamine amidohydrolase | |
Mouse | |
65 kDa | |
0.1 mL | |
Primary | |
Specific for GAC but not KGA isoform. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction