Learn More
Mouse anti-Glutamine Synthetase, Clone: 7711, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243826
Catalog No. NBP243826
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Glutamine Synthetase
7711
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000
Purified
GLUL
Full length human Glutamine synthetase Recombinant protein.
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
PBS with No Preservative
cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Mouse
IgG1
0.1 ml
Hypoxia, Neurodegeneration, Neuroscience, Vision
Primary
2752