Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Glutathione S Transferase kappa 1, Clone: 16A11.E4.E3.E7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244172

Catalog No. NBP244172


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page

Glutathione S Transferase kappa 1 Monoclonal antibody specifically detects Glutathione S Transferase kappa 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page.
Specifications

Specifications

Glutathione S Transferase kappa 1
16A11.E4.E3.E7
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
EC 2.5.1.18, glutathione S-transferase k1, glutathione S-transferase kappa 1, Glutathione S-transferase subunit 13, glutathione S-transferase subunit 13 homolog, GST, GST 13-13, GST class-kappa, GST13, GST13-13, GSTK1-1, hGSTK1
Mouse
IgG2a Lambda
0.1 mg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page
1 mg/mL
Western Blot 1:500 - 1:1000, ELISA 1:50000 - 1:200000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, SDS-Page 1:100-1:2000
Purified
GSTK1
This Protein A purified antibody was prepared by repeated immunizations in mice with Glutathione S Transferase kappa 1 Protein.
Protein A purified
RUO
Primary
373156
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only