Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Goosecoid, Clone: 4C5D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237356

Catalog No. NBP237356


Add to cart

Description

Description

Goosecoid Monoclonal antibody specifically detects Goosecoid in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Goosecoid
4C5D10
Unconjugated
PBS with 0.05% Sodium Azide
goosecoid homeobox, homeobox protein goosecoid
Mouse
IgG1
Protein A or G purified
RUO
Primary
145258
Western Blot, ELISA
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
Affinity Purified
GSC
Purified recombinant fragment of human GSC (AA: 191-257) expressed in E. Coli.
28.2 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only