Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPBB, Mouse, Clone: 17B6, Abnova™

Mouse monoclonal antibody raised against GPBB.

Manufacturer:  Abnova Corporation MAB2956

Catalog No. 89-135-317


Add to cart

Description

Description

Specifications

Specifications

GPBB
17B6
Mouse monoclonal antibody raised against GPBB.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG1
RUO
Primary
This antibody reacts with BB and MM isoenzymes.
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Human GPBB.
200 ug
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit