Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth hormone, Mouse, Clone: YC6, Abnova™

Catalog No. 89136417 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-417 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-417 Supplier Abnova Corporation Supplier No. MAB8115
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Growth hormone.

Specifications

Antigen Growth hormone
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone YC6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Growth hormone.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Host Species Mouse
Immunogen Recombinant protein corresponding to human Growth hormone.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.