Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSR, Mouse, Clone: 1A7, Abnova™

Mouse monoclonal antibody raised against partial recombinant GSR.

Manufacturer:  Abnova Corporation MAB0693

Catalog No. 89-108-049


Add to cart

Description

Description

Specifications

Specifications

GSR
1A7
Mouse monoclonal antibody raised against partial recombinant GSR.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
GSR
Recombinant protein corresponding to amino acids 44-522 of human GSR.
Ammonium sulfate precipitation
RUO
Primary
2936
ELISA, Immunoprecipitation
Unconjugated
Immunoprecipitation (1-2 ul) The optimal working dilution should be determined by the end user.
GSR
MGC78522
Mouse
IgG2a, kappa
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit