Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA tag, Mouse, Clone: HA.C5/C10, Abnova™

Catalog No. 89136773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-773 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-773 Supplier Abnova Corporation Supplier No. MAB9751
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of HA tag.

Specifications

Antigen HA tag
Applications Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HA.C5
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of HA tag.
Dilution Western Blot (1:1000-1:3000) Immunohistochemistry (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.05% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to HA tag.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the N-terminal, C-terminal or internal HA- tagged fusion proteins.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.