Learn More
Cell Biolabs IncSupplier Diversity Partner MOUSE ANTI-HA TAG MONOCLON AB  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner AKR006
Mouse Anti-HA Tag Monoclonal Antibody is Protein A purified and provided at 1 mg/mL. Suitable for Western blot or Immunostaining. 100 ug.
Catalog No. 50-672-569