Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hep Par-1, Mouse, Clone: OCH1E5, Abnova™

Mouse monoclonal antibody raised against Hep Par-1.

Manufacturer:  Abnova Corporation MAB7927

Catalog No. 89-136-400

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Hep Par-1
OCH1E5
Mouse monoclonal antibody raised against Hep Par-1.
Liquid concentrate in PBS (BSA, 0.09% sodium azide)
Mouse
IgG1
RUO
Primary
Hepatocellular antigen of the mitochondrial fraction.
Immunohistochemistry
Unconjugated
Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Liquid
Formalin-fixed, rejected liver transplant.
1 mL
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit