Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B virus surface antigen, Mouse, Clone: DNH002, Abnova™

Mouse monoclonal antibody raised against full-length recombinant hepatitis B virus surface antigen.

Manufacturer:  Abnova Corporation MAB1690

Catalog No. 89-135-015

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Sequence: virus
Specifications

Specifications

Hepatitis B virus surface antigen
DNH002
Mouse monoclonal antibody raised against full length recombinant hepatitis B virus surface antigen.
In 10mM phosphate buffer, 137mM NaCl, 2.7mM KCl, pH 7.4 (0.09% sodium azide)
Mouse
IgG1
RUO
Primary
This antibody is specific against HBsAg Ad.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to full length hepatitis B virus surface antigen.
1 mg
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit