Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus core antigen, Mouse, Clone: B055M, Abnova™

Mouse monoclonal antibody raised against recombinant hepatitis C virus core antigen.

Manufacturer:  Abnova Corporation MAB4784

Catalog No. 89-135-784

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Hepatitis C virus core antigen
B055M
Mouse monoclonal antibody raised against recombinant hepatitis C virus core antigen.
In 10mM PBS, pH 7.2 (0.09% sodium azide)
Mouse
IgG2a
RUO
Primary
Hepatitis C Virus (HCV) Core Antigen. Reactive with genotypes 1a and 2. Does not work in Western blot or IFA.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to hepatitis C virus core antigen (genotype 1a).
1 mg
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit