Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus core protein, Mouse, Clone: H6-29, Abnova™

Mouse monoclonal antibody raised against recombinant hepatitis C virus core protein.

Manufacturer:  Abnova Corporation MAB7949

Catalog No. 89-136-406


Add to cart

Description

Description

Specifications

Specifications

Hepatitis C virus core protein
H6-29
Mouse monoclonal antibody raised against recombinant hepatitis C virus core protein.
In PBS (50% glycerol)
Mouse
IgG2a, kappa
RUO
Primary
This antibody is specific to human HCV core antigen.
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to amino acids 13-124 of HCV core protein.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit