Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Histone H3, Biotin, Clone: 1B1-B2, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236468B

Catalog No. NBP236468B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Histone H3
1B1-B2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
HIST2H3C
Synthetic peptide corresponding to the C-terminus of human Histone H3.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
PBS with 0.05% Sodium Azide
H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Mouse
IgG3 Kappa
0.1 ml
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Phospho Specific
Primary
126961
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit